Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Telluric acid dihydrate, 99+%

Catalog No. p-4904548
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1419709 10 g
AA1419718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1419709 Supplier Thermo Scientific Chemicals Supplier No. 01419709
Only null left
Add to Cart

CAS: 13451-14-4 | H6O6Te | 229.64 g/mol

Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13451-14-4
Molecular Formula H6O6Te
Molecular Weight (g/mol) 229.64
MDL Number MFCD00150387
InChI Key VCYUGZHEVVXOPD-UHFFFAOYSA-N
Synonym telluric acid dihydrate, telluricacidwhitepowder, telluric acid-te

Specifications

Boiling Point 160°C (decomposition)
Melting Point 136°C
Color White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range >99%
Solubility Information Soluble in water.
Formula Weight 229.64 (193.61 Anhydrous)
Sensitivity Hygroscopic
Density 3.158
Chemical Name or Material Telluric acid dihydrate
Show More Show Less
Hazard Category H315-H319-H332-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 232-267-6
RTECSNumber WY2350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.