Learn More
TARGETMOL CHEMICALS INC TELOMERASE-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T12009
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telomerase-IN-1 is a telomerase inhibitor (IC50 0.19 uM). telomerase-IN-1 activates hTERT expression through the endoplasmic reticulum stress (ERS) of the ER overreaction (EOR) which then induces ERS leading to apoptosis. telomerase-IN-1 leads to inhibition of cell proliferation through the expression of downstream signaling molecules including Telomease-IN-1 has high inhibitory activity against telomerase high anti-proliferative capacity against SMMC-7721 cells and no significant toxic effect on human normal hepatocytes. telomerase-IN-1 in xenograft tumor model exhibited antitumor activity. purity: 97%
Catalog No. 50-258-1491
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.