Description
- Manufactured with PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) solvents
- PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) are ideal for polar water-soluble analyte extraction from aqueous samples.
- PS-DVB ENV has a very high surface area, offering excellent retention of very polar analytes including phenols and phenoxyacid herbicides from environmental samples and drug metabolites from biological fluids.
- Columns are made of polypropylene and feature Luer-slip outlet.
Specifications
Specifications
| Bed Weight | 200 mg |
| For Use With (Application) | Polar Water-soluble Analyte Extraction |
| For Use With (Equipment) | Conventional HPLC System |
| Sorbent | PS-DVB (ENV) |
| Type | Solid Phase Extraction Column |
| Volume (Metric) | 6 mL |