Learn More
Electron Microscopy Sciences Omniprobe TEM Grid & Sample Holder, Stainless steel
SDP
Supplier:  Electron Microscopy Sciences 75969SS
Encompass_Preferred
TEM grid holder with stations for 2 TEM grids and 2 sample stubs. Long post – standard is 1/8" (3.2 mm) diameter x 0.32" (8.1 mm) length. Comes with 2 sample stubs. Stainless Steel is very slightly magnetic.
Catalog No. 50-337-75
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.