Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TEM7/PLXDC1 Antibody Pair [HRP]
SDP

Catalog No. NB009765 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009765 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009765 Supplier Novus Biologicals™ Supplier No. NBP27936315PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 156.25 pg/mL
Assay Sensitivity 156.25-10000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TEM7/PLXDC1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7
Gene ID (Entrez) 57125
Gene Symbol PLXDC1
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-TEM7/PLXDC1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.3 mg/mL of mouse anti-TEM7/PLXDC1 monoclonal antibody.
  • Standard: Each vial contains 270 ng of recombinant TEM7/PLXDC1.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Angiogenesis
    Storage Requirements Storage is content dependent.
    Target TEM7/PLXDC1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.