Learn More
Cayman Chemical Temozolomide, 85622-93-1, 50 mg
SDP
Supplier:  Cayman Chemical 1416350
Encompass_Preferred
An imidazotetrazine that is converted to a compound capable of alkylating DNA, thus interfering with DNA replication and leading to cytotoxicity in proliferating cells; rapidly and completely absorbed from the gastrointestinal tract after oral administration and readily crosses the blood-brain barrier
Catalog No. NC0582294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.