Learn More
Thermco Products Temp/Humidity/Dew Point Logger
Catalog No. 04500339
Change view
Click to view available options
Display Type:
LCD
Catalog No. Display Type
04-500-339 LCD
1 options
Catalog No. 04-500-339 Supplier Thermco Products Supplier No. ELWIFITH
Add to Cart
Add to Cart
This WiFi-enabled data logger monitors temperature, humidity, and dew point.
  • Measurement ranges include -20°C to 60°C (-4°F to 140°F) and 0 to 100% RH.
  • Transfers data to the EasyLog Cloud via WiFi or to a host PC for real-time tracking.
  • Allows remote customization of settings, email alerts for alarms, linking multiple sensors to one account, and assignment of user privileges.
  • If WiFi connectivity is lost, data is stored internally and automatically synchronized to the Cloud upon reconnection.
  • EasyLog Cloud Apps for Android and Apple simplify setup and provide convenient data access.
  • Provides comprehensive monitoring of temperature, humidity, and dew point.
  • Uses standard WiFi for seamless data transfer and connectivity.
  • Ensures data resilience with internal storage during WiFi outages.
  • Complies with BS EN 12830:2018 requirements for temperature-sensitive goods.
Specifications
Type Temperature and Humidity Datalogger
Display Type LCD
Alarm Types E-mail