Learn More
BINDER™ Temperature Calibration Certificate
Temperature measurement and certificate, measured at specified temperature.
Supplier: BINDER™ 80122425
Description
- Calibration measurement taken at specified temperature.
- Choose between 9, 15 to 18, 27, 28, 29, or 30 measurement points
Specifications
| KB PRO Series | |
| 28 Measuring Points | |
| 1 Day | |
| Calibration Certificate |