Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Template Switching Reverse Transcription Oligonucleotides

Catalog No. p-200009446
Encompass_Preferred
Change view
Click to view available options
Description:
Capturing Oligo (dT) Primer
Preamplification Primer
Template Switching Oligo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
A65427 Template Switching Oligo
A65428 Preamplification Primer
A65429 Capturing Oligo (dT) Primer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A65427 Supplier Invitrogen™ Supplier No. A65427
Only null left
Add to Cart
Edge
Add to Cart

Use this set of oligonucleotides for template switching cDNA synthesis and preamplification. Template Switching Oligonucleotide and Capturing Oligo (dT) are used to incorporate adaptor sequences to full-length cDNA. Preamplification primer enables PCR amplification of obtained cDNA.

This set of oligonucleotides is designed for template switching reverse transcription reaction and following preamplification of obtained cDNA. Template Switching Oligonucleotide (TSO) and Capturing Oligo (dT) are used during cDNA synthesis reaction to incorporate adaptor sequences in both ends of full-length cDNA. Preamplification primer is complementary to the adaptor sequence and enables PCR amplification of obtained cDNA.

Template Switching Oligonucleotide (TSO) and Capturing Oligo (dT) Primer are designed for use in template switching reverse transcription reaction. Capturing oligonucleotide containing oligo(dT) and adapter sequences is used as a primer for selective first-strand cDNA synthesis from poly(A)-containing RNAs. When the reverse transcriptase reaches the 5ʹ end of the RNA template, terminal deoxynucleotidyl transferase (TdT) activity of reverse transcriptase adds 1–3 extra nucleotides to the cDNA end, enabling hybridization of the template switching oligo and ligation-free incorporation of adapter sequence to the 3ʹ end of the resulting cDNA. Both oligos are supplied as ready-to-use 20X concentrated solutions. For maximal template switching efficiency, we recommend using Template Switching Oligo and Capturing oligo (dT) Primer with Invitrogen SuperScript IV Template Switching RT Master Mix .

Adapter sequences incorporated at both ends of the cDNA may serve as primer-binding sites in the subsequent PCR amplification step, allowing global preamplification of full-length templates. For PCR, use Preamplification Primer which is complementary to TSO and Capturing Oligo (dT) adaptor sequence and is supplied as ready-to-use 100X concentrated solution.

The TSO, Capturing Oligo (dT) and Preamplification primers are also available as part of complete Invitrogen SuperScript IV Single Cell/Low Input cDNA PreAmp Kit (Cat. No. 11752048) .

Applications
• cDNA synthesis
• cDNA preamplification
• RNA-seq
• RACE

TRUSTED_SUSTAINABILITY

Specifications

Description Template Switching Oligo
Product Type Oligonucleotide
Content And Storage Template Switching Oligo, 48 μL

Store at -25°C to -15°C.
Concentration 20X
Label or Dye No
Quantity 48 Reactions
Color Colorless
For Use With (Application) cDNA synthesis

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.