Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva TempliPhi™ Sequence Resolver Kit
GREENER_CHOICE

Catalog No. 45002402 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
20 reactions
200 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-002-402 20 reactions
45-002-266 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-402 Supplier Cytiva Supplier No. 28903529
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Sample buffer, Reaction buffer and Enzyme mix

Uses modified nucleotides in isothermal rolling circle amplification (RCA) to improve results with difficult to sequence templates

  • Cultureless preparation from bacterial colonies, glycerol stock or purified DNA, saves time and reagents
  • Requires 20 minutes hands-on time
  • Excellent sequence finishing performance resolving sequencing stop and compression issues associated with difficult templates
  • Improve difficult template sequencing success with up to 820 bp Phred 20 read length
  • For use directly in cycle sequencing reactions without purification
  • Available in sizes of 20, 50 and 500 reactions

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°CThaw components on ice and maintain at 0°C to 4°C during handling
Description illustra TempliPhi Sequence Resolver Kit uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing.
Format Kit
For Use With (Application) Amplification of difficult templates for successful DNA sequencing
Includes 1 x 90μL Sample Buffer, 1 x 90μL Reaction Buffer, 1 x 90μL Enzyme mix
No. of Reactions 20 reactions
Product Type illustra TempliPhi Sequence Resolver Kit
Test Time 18 h
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.