Learn More
Abcam TEMPO, Stable Nitroxyl Radical, Solid, 500 mg
Description
- Molecular weight: 156.25 g/mol
- Purity: >98%
- Form: Solid
- Applications: Radical trap, structural probe for biological systems in electron spin resonance spectroscopy, and catalyst for the oxidation of primary alcohols to aldehydes
- Storage buffer: 100% TEMPO
- Storage temperature: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | TEMPO |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Quantity | 500 mg |
| For Use With (Application) | Radical trap, structural probe for biological systems, electron spin resonance spectroscopy, catalyst for the oxidation of primary alcohols to aldehydes, carboxylation of water-resistant nanofibrillated cellulose films |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.