Learn More
Enzo Life Sciences TEMPONE-H. hydrochloride (10mg). CAS: 3637-11-4
SDP
Supplier:  Enzo Life Sciences ALX430071M010
Encompass_Preferred
Very effective, cell permeable and non-toxic spin trap for the detection of superoxide radical and peroxynitrite. Alternative name: 1-Hydroxy-2,2,6,6-tetramethyl-4-oxo-piperidine . HCl. Purity: ≥99% (GLC). Appearance: White to off-white crystalline powder. Solubility: Soluble in DMSO, water, methanol or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9181
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.