Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 6 SFR Hooded Coverall

Catalog No. 191307802 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
Blue
Light Blue
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-130-7802 Light Blue 2X-Large
19-175-272 Blue 6X-Large
19-100-386 Blue 5X-Large
19-054-006 Blue 4X-Large
19-130-7801 Light Blue X-Large
19-130-7800 Light Blue Large
19-130-7803 Light Blue 3X-Large
19-175-270 Blue Medium
19-175-273 Blue 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Should not be worn in presence of hazardous materials.
Catalog No. 19-130-7802 Supplier DuPont™ Supplier No. TM127SBU2X002500
Add to Cart
Add to Cart
This item is not returnable. View return policy
Should not be worn in presence of hazardous materials.

Offer an extra layer of protection for primary fire-resistant clothing

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.

Compliance

Meets ANSI™/ISEA 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standards Methods of Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.

TRUSTED_SUSTAINABILITY

Specifications

Size 2X-Large
Material ProShield
Color Light Blue
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper
Seam Type Serged
Description Non-woven Wood Pulp, Polyester Fabric
Size (English) Chest 44.25 to 47.75 in.
Size (English) Inseam 30.5 in.
Size (Metric) Chest 112.39 to 121.28 cm
Size (Metric) Inseam 77.47 cm
For Use With (Application) Steel mills, Coal mines, maintenance work, refineries, laboratories, utilities
Length (English) Sleeve 38.25 in.
Length (Metric) Sleeve 97.15 cm
Width (English) Chest 28.75 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.