Learn More
Rudolph Research Analytical Rudolph Research TempTrol 100MM Ceramic Cell, Alumina Material, High Corrosion Resistance, 2.0 mL
SDP
Supplier:  Rudolph Research Analytical 60T5.01002CU
Encompass
The TempTrol 100MM Ceramic Cell is a high-performance sample cell designed for accurate polarimetry, featuring excellent chemical stability and heat resistance. With its robust alumina ceramic construction and a pathlength of 100 mm, it ensures precise temperature control and compatibility with Rudolph Research Polarimeters, enhancing sample analysis by minimizing air bubbles. Its biocompatibility and corrosion resistance set it apart in demanding scientific applications.
Catalog No. NC2186683
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.