Learn More
Medchemexpress LLC Tenacissoside I | 191729-44-9 | MFCD22417327 | 99.9% | 814.95 | C44H62O14 | 10 MG
Supplier: Medchemexpress LLC HYN210210MG
Tenacissoside I is a C21 steroidal glycoside isolated from the plant Marsdenia tenacissima. It is supplied as a high-purity research reagent for natural-product, analytical, and pharmacological studies.
- High purity: 99.93%.
- Natural C21 steroidal glycoside from Marsdenia tenacissima.
- Molecular weight 814.95 and formula C44H62O14.
- Available in small quantities suitable for analytical and biochemical research.
- Includes datasheet, certificate of analysis, and safety data sheet.