Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Foam Nitrile Coating
Catalog No. 17401240
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-240 6
17-401-239 5
17-401-241 7
17-401-242 8
17-401-244 10
17-401-245 11
17-401-243 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-401-240 Supplier Superior Glove™ Supplier No. S13TAFNT6
Add to Cart
Add to Cart
This item is not returnable. View return policy
Knit with high-tenacity composite yarns designed to meet a range of applications. Foam nitrile coating on palm provides secure grip on wet or oily parts.
  • Continuous filament yarn is stronger than steel on an equal-weight basis
  • 1250 grams of cut protection
  • Lint-free
Nitrile Palm Coat Style
  • Withstands oils and solvents
  • Touch screen compatible
  • Black and Gray
  • Ideal for handling sharp, oily glass or metal
  • ANSI Cut Level A4
  • High coefficient of friction
  • Non-shedding
Specifications
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A3
Material Composite Fiber
Product Type Cut-resistant Gloves
Size 6
For Use With (Application) Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
Glove Cuff Style Knit