Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Foam Nitrile Coating
Description
- Continuous filament yarn is stronger than steel on an equal-weight basis
- 1250 grams of cut protection
- Lint-free
Nitrile Palm Coat Style
- Withstands oils and solvents
- Touch screen compatible
- Black and Gray
- Ideal for handling sharp, oily glass or metal
- ANSI Cut Level A4
- High coefficient of friction
- Non-shedding
Specifications
Specifications
| Coating Material | Nitrile Foam |
| Color | Black |
| Cut Resistance Rating | ANSI A3 |
| Material | Composite Fiber |
| Product Type | Cut-resistant Gloves |
| Size | 9 |
| For Use With (Application) | Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |
| Glove Cuff Style | Knit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.