Learn More
Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Polyurethane Coating
Catalog No. 17401251
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-251 9
17-401-247 5
17-401-248 6
17-401-249 7
17-401-250 8
17-401-252 10
17-401-253 11
17-401-254 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-401-251 Supplier Superior Glove™ Supplier No. S13TAGPU9
Add to Cart
Add to Cart
This item is not returnable. View return policy
Knit with high-tenacity composite yarns designed to meet a range of applications. Polyurethane coating offers a high coefficient of friction that is non-shedding.
  • Continuous filament yarn is stronger than steel on an equal-weight basis
  • 1250 grams of cut protection
  • Lint-free
Withstands oils and solvents
  • Touch screen compatible
  • Gray
  • Ideal for handling sharp, oily glass or metal
  • ANSI Cut Level A2
  • Polyurethane-Coated Style
    • Non-shedding
    • High coefficient of friction
    Specifications
    Coating Material Polyurethane
    Color Gray
    Cut Resistance Rating ANSI A2
    Material Composite Fiber
    Product Type Cut-resistant Gloves
    Size 9
    For Use With (Application) Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
    Glove Cuff Style Knit