Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ TenActiv™ Touchscreen Compatible Static and Cut-Resistant Gloves

Catalog No. 17400201
Change view
Click to view available options
Size:
0
1
2
5
6
7
8
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-201 0
17-400-202 1
17-400-203 2
17-400-197 6
17-400-198 7
17-400-199 8
17-400-200 9
17-400-196 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-400-201 Supplier Superior Glove™ Supplier No. S13TAPUCF0
Add to Cart
Add to Cart

Cut-resistant gloves with polyurethane coating on palms and fingers, as well as touchscreen compatibility.

Perfect for electronics manufacturing, these gloves are made from a 13-gauge TenActiv™ blended yarn that provides ANSI Level A2 cut protection.

  • Carbon filament imparts anti-static and electrostatic dissipating (ESD) properties
  • Polyurethane palm coating for a strong, steady grip
  • Free of DMF (dimethylformamide)
  • Touchscreen compatible to move seamlessly between tasks

Specifications

Coating Material Polyurethane
Color Gray, Yellow
Cut Resistance Rating ANSI A2
Material TenActiv Blended Yarn
Size 0
For Use With (Application) Electronics and Glass Manufacturing, Automotive Industry
Puncture Rating ANSI/ISEA 105-16 Level 3
Yarn Gauge 13
Length (English) Cuff 2 in.
Length (Metric) Cuff 5.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.