Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sempermed™ Tender Touch™ 100 Nitrile Gloves

Catalog No. 19048618 Shop All Sempermed USA Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-618 Medium
19-048-616 X-Small
19-048-617 Small
19-048-619 Large
19-048-621 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-048-618 Supplier Sempermed™ Supplier No. TTNF103
This item is not returnable. View return policy

Powder free textured nitrile gloves are ideal for medical, dental, veterinarian and laboratory applications.

  • 100 gloves per box (all sizes)
  • Chemical resistant
  • Enhanced protection
  • Manufactured without natural rubber
  • Tested for use with chemotherapy drugs
  • Meets USP 800 requirements for PPE compliance for chemotherapy

Specifications

Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory
AQL 1
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 2.8 mil
Finger Thickness (Metric) 0.07 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.4 mil
Palm Thickness (Metric) 0.06 mm
Size Medium
FDA 510 (k) Yes
Model TTNF103
Series Tender Touch™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.