Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Teniposide
SDP

Catalog No. 6975100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-751-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 69-751-00 Supplier Tocris Bioscience™ Supplier No. 6975/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DNA topoisomerase II inhibitor; viral transduction enhancer

Teniposide is a DNA topoisomerase II inhibitor and viral transduction enhancer. It induces breaks in double-stranded DNA and prevents DNA repair. Teniposide enhances transduction of recombinant adeno-associated virus 2 (rAAV2) by up to 40-fold in human cells in vitro and by 100-fold in vivo at 24 h post-transduction.

Specifications

Chemical Name or Material (5 R,5 aR,8 aR,9 S)-5,8,8 A,9-Tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-9-[[4,6-O-[(R)-2-thienylmethylene]-β-D-glucopyranosyl]oxy]furo[3 ',4 ':6,7 ]naphtho[2,3-d]-1,3-dioxol-6(5 aH)-one
CAS 29767-20-2
Quantity 100 mg
Target DNA Topoisomerase Inhibitors
Molecular Formula C32H32O13S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.