Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TENM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590694 Supplier Invitrogen™ Supplier No. PA590694
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPVEGELYAN GVSKGNRGTE SMDTTYSPIG GKVSDKSEKK VFQKGRAIDT GEVDIGAQVM QTIPPGLFWR FQITIHHPIY LKFNISLAKD SLLGIYGRRN IPPTHTQFDF VKLMDGKQLV KQDSKGSDDT QHSPRNLILT SLQETGFIEY MDQGPWYLAF YNDGKKMEQV FVLTTAIEIM DDCSTNCNGN G; Positive Samples: U-87MG, U-251MG, HeLa; Cellular Location: Cell membrane, Cytoplasm, Nucleus, Nucleus matrix, Nucleus speckle, Single-pass membrane protein, cytoskeleton.

The teneurin transmembrane protein 1 (TENM1) is a member of a family of four neuronal cell surface proteins homologous to the Drosophila pair-rule gene Ten-m. TENM1 is expressed primarily in the developing central nervous system and may be proteolytically cleaved with the intracellular domain translocating to the nucleus. TENM1 is a direct target of the homeobox transcription factor EMX2, a transcription factor thought to be important for area specification in the developing cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TENM1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TENM1
Gene Accession No. Q9UKZ4, Q9WTS4
Gene Alias IDten-1; odz (odd Oz/ten-m, Drosophila) homolog 3; odz, odd Oz/ten-m homolog 1; odz, odd Oz/ten-m homolog 1(Drosophila); Odz1; Odz2; ODZ3; Protein Odd Oz/ten-m homolog 1; RGD1561678; TCAP-1; TCPA-1; Ten-1; Ten-1 ECD; Ten-1 extracellular domain; Ten-1 intracellular domain; tenascin M1; tenascin-M1; teneurin 1; Teneurin C-terminal-associated peptide; Teneurin transmembrane protein 1; teneurin-1; TENM1; Ten-m1; TNM; TNM1
Gene Symbols TENM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-540 of human TENM1 (NP_0011567501).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10178, 23963, 298168
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.