Learn More
Apexbio Technology LLC Tenofovir,25mg CAS# 147127-20-6
SDP
Supplier:  Apexbio Technology LLC A527525
Encompass_Preferred
Selectively inhibits HIV reverse transcriptase (RNA-dependent DNA polymerase). Prevents cytotoxicity in SIV-infected C-8166 cells in vitro (IC50 = 1.5 μM). Antiviral agent. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0466
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.