Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tenovin-1, p53 Activator, >98% Purity, 10 mg

Catalog No. 502707165
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7165 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7165 Supplier Abcam Supplier No. AB12088910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent p53 activator that protects p53 from MDM2-mediated degradation through SirT1 inhibition, exhibiting antiproliferative effects in vitro.

  • Purity: >98%
  • Molecular Weight: 369.48 g/mol
  • Mechanism: p53 activator; protects p53 from MDM2-mediated degradation via SirT1 inhibition
  • Biological Effects: Inhibits downstream histone acetylation, activates p21CIP/WAF1, and demonstrates potent antiproliferative effects in vitro
  • Storage: Store at +4°C
  • Composition: 100% Tenovin-1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule p53, SirT1, p21CIP/WAF1
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Buffer 100% Tenovin-1
Content And Storage Store at 4°C.
Formulation 100% Tenovin-1
Quantity 10 mg
For Use With (Application) p53 activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.