Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Tenovin-1, p53 Activator, >98% Purity, 50 mg

Catalog No. 502707164
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7164 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7164 Supplier Abcam Supplier No. AB12088950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small molecule p53 activator that protects p53 from MDM2 mediated degradation through SirT1 inhibition.

  • Molecular weight: 369.48 g/mol
  • Purity: >98%
  • Description: p53 activator that protects p53 from MDM2 mediated degradation through SirT1 inhibition
  • Biological activity: Inhibits downstream histone acetylation and activates p21CIP/WAF1
  • Application: Potent antiproliferative effects in vitro

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule p53, SirT1, p21CIP/WAF1
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Buffer 100% Tenovin-1
Content And Storage Store at 4°C.
Form Solid
Formulation 100% Tenovin-1
Quantity 50 mg
For Use With (Application) p53 activation, histone acetylation inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.