Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tensin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557515 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557515 Supplier Invitrogen™ Supplier No. PA557515
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: LSPLTNGLDK SYPMEPMVNG GGYPYESASR AGPAHAGHTA PMRPSYSAQE GLAGYQREGP HPAWPQPVTT SHYAHDPSGM FRSQS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tensin 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNS1
Gene Accession No. Q9HBL0
Gene Alias 1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1
Gene Symbols TNS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Tensin 1. Recombinant protein control fragment (Product #RP-96508).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.