Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TER-119/Erythroid cells Rat anti-Mouse, BV421, Clone: TER-119, BD Horizon™

Catalog No. BDB566248 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566248 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566248 Supplier BD Biosciences Supplier No. 566248
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

TER-119/Erythroid cells Monoclonal antibody specifically detects TER-119/Erythroid cells in Mouse samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TER-119/Erythroid cells
Applications Flow Cytometry
Classification Monoclonal
Clone TER-119
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119
Host Species Rat
Immunogen Mouse Fetal Liver
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.