Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbufos, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
13071-79-9,67-56-1
C9H21O2PS3
288.419
XLNZEKHULJKQBA-UHFFFAOYSA-N
25670
CHEBI:38960
tert-butylsulfanylmethylsulfanyl-diethoxy-sulfanylidene-$l^{5}-phosphane
CCOP(=S)(OCC)SCSC(C)(C)C
13071-79-9,67-56-1
C9H21O2PS3
288.419
XLNZEKHULJKQBA-UHFFFAOYSA-N
25670
CHEBI:38960
tert-butylsulfanylmethylsulfanyl-diethoxy-sulfanylidene-$l^{5}-phosphane
CCOP(=S)(OCC)SCSC(C)(C)C

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Terbufos13071-79-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
CAS Min % 0.1,99.9
CAS Max % 0.1,99.9
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Solvent or Matrix Methanol
Quantity 1mL
Chemical Name or Material Terbufos

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only