Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Terbutaline Hemisulfate, Beta2 Receptor Agonist, Solid, 1 g

Catalog No. 502707260
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7260 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7260 Supplier Abcam Supplier No. AB1207921G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective β2 receptor agonist with >99% purity that induces cAMP production.

  • Purity: >99%
  • Mechanism: Selective β2 receptor agonist (EC50 = 2.3 μM)
  • Activity: Full agonist induces cAMP production
  • Molecular Weight: 274.32 g/mol
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Beta2 Adrenergic Receptor
Form Solid
Content And Storage Store at ambient temperature.
Quantity 1 g
For Use With (Application) cAMP production
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.