Learn More
Abcam Terbutaline Hemisulfate, Beta2 Receptor Agonist, Solid, 5 g
Description
- Molecular weight: 274.32 g/mol
- Purity: >99%
- Mechanism of action: Selective β2 receptor agonist (EC50 = 2.3 μM)
- Functional activity: Full agonist induces cAMP production
- Form: Solid
- Composition: 100% Terbutaline sulphate
- Storage: Store at Ambient
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Beta2 Adrenergic Receptor |
| Buffer | 100% Terbutaline sulphate |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Quantity | 5 g |
| For Use With (Application) | Beta2 receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.