Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthalbis(p-phenetidine) 98.0+%, TCI America™
SDP

Catalog No. B087125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B087125G 25 g
B08715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B087125G Supplier TCI America Supplier No. B087125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17696-60-5
Molecular Formula C24H24N2O2
Molecular Weight (g/mol) 372.468
MDL Number MFCD00026796
InChI Key KSHALPAIUOUGBA-UHFFFAOYSA-N
Synonym Bis(p-ethoxyanilino)terephthalaldehyde, Bisphenetidylterephthalaldehyde, Di-p-phenetidylterephthalaldehyde
PubChem CID 632961
IUPAC Name N-(4-ethoxyphenyl)-1-[4-[(4-ethoxyphenyl)iminomethyl]phenyl]methanimine
SMILES CCOC1=CC=C(C=C1)N=CC2=CC=C(C=C2)C=NC3=CC=C(C=C3)OCC

Specifications

Quantity 25 g
Formula Weight 372.47
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Terephthalbis(p-phenetidine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.