Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco TERi Androgynous Geriatric Patient Care Trainer - Light
SDPSCI_ED

Catalog No. S97908 Shop All Simulaids, Inc. Products
Change view
Click to view available options
Color:
Light
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S97908 Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S97908 Supplier Nasco Supplier No. LF04300
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

A comprehensive male/female elderly patient care trainer for medical simulation. Evolving from our GERi/KERi full body manikins, it has a more realistic look and feel accurately representing the human anatomy. Weight is distributed to represent a real patient for lifting and carrying.

Passive Range of Motion

  • Seating, laying, finger/toe manipulation with no effort from patient.

Enhanced Aesthetics

  • Soft silicone skin with noticeable normal and cancerous skin moles, stage 1 sacral ulcer and reddened skin folds.

Grooming and Daily Living Assistance

  • Bathing, clothing changes, denture placement/removal, hearing aid placement/removal, eye irrigation and optic drops, ostomy and urinary catheter care, hair care.

Specifications

Product Type Geriatric Patient Care Trainer
For Use With (Application) Simulating Patient Care
Color Light
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.