Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco TERi Androgynous Geriatric Patient Skills Trainer - Light
SDPSCI_ED

Catalog No. S97909 Shop All Simulaids, Inc. Products
Change view
Click to view available options
Color:
Light
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S97909 Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S97909 Supplier Nasco Supplier No. LF04301
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

A comprehensive male/female elderly patient care trainer for medical simulation. Evolving from our GERi/KERi full body manikins, it has a more realistic look and feel accurately representing the human anatomy. Weight is distributed to represent a real patient for lifting and carrying.

Passive Range of Motion

  • Seating, laying, finger/toe manipulation with no effort from patient.

Cardiovascular Support

  • Carotid pulses, CPR with quality performance metrics and auscultation.

Intravenous, Injection, Subcutaneous Injection Sites

  • Multiple locations equipped with standard arm IM injection pads.

Nasogastric Tube Procedures

  • Placement, lavage, gavage, feeding and suctioning.

Enhanced Aesthetics

  • Soft silicone skin with noticeable normal and cancerous skin moles, stage 1 sacral ulcer and reddened skin folds.

Respiratory Support

  • Manual ventilation with chest rise, oxygen administration, tracheostomy care, airway/trach suctioning. Airway intubation, ventilation and auscultation.

Grooming and Daily Living Assistance

  • Bathing, clothing changes, denture placement/removal, hearing aid placement/removal, eye irrigation and optic drops, ostomy and urinary catheter care, hair care.

Indwelling Male and Female Urinary Catheterization

  • Includes support for urethral and suprapubic catheters.

Advanced Skills

  • Urinary catheterization, enema administration, pelvic and prostate examination.

Specifications

Product Type Geriatric Patient Skills Trainer
For Use With (Application) Simulating Patient Care
Color Light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.