Learn More
Medchemexpress LLC HY-P0059 100mg , Teriparatide PTH 1-34;hPTH (1-34);Human parathyroid hormone-(1-34) CAS:52232-67-4 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYP0059100MG
Encompass_Preferred
Medchemexpress, HY-P0059 100mg Teriparatide PTH 1-34;hPTH (1-34);Human parathyroid hormone-(1-34) CAS:52232-67-4 Formula:C181H291N55O51S2 IC50: 2 nM (PTH) . Purity:98% Teriparatide is a PHT agonist, with an IC 50 of 2 nM in HEK293 cells. Sequence: Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3351
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.