Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terminal deoxynucleotidyl transferase (TdT), Murine, 2500 U, G-Biosciences
SCI_ED

Catalog No. S001115
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001115 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001115 Supplier G-Biosciences Supplier No. 7862260
Add to Cart
Add to Cart

Terminal Transferase catalyzes the untemplated addition of deoxynucleotides to the 3' hydroxyl terminus of DNA molecules.

  • This enzyme is derived from the dntt gene from Mus Musculus, and is the Isoform S version, which is a 131 amino acid N-terminal truncation of the enzyme described in Comparison of the Two Murine Deoxynucleotidyltransferase Terminal Isoforms, Boule J.-B., Rougeon F., Papanicolaou C.J. Biol. Chem. 275:28984-28988 (2000)

Specifications

Packaging Vial
Enzyme Terminal Deoxynucleotidyl Transferase
Sensitive to Heat Inactivation Yes
Product Type Terminal Transferase
Research Category DNA Repair, RNA Research
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.