Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terpinyl Acetate 85.0+%, TCI America™
SDP

Catalog No. T002325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T002325ML 25 mL
T0023500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T002325ML Supplier TCI America Supplier No. T002325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80-26-2
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00037155
InChI Key IGODOXYLBBXFDW-UHFFFAOYNA-N
Synonym alpha-terpinyl acetate, terpineol acetate, terpineol, acetate, p-menth-1-en-8-yl acetate, p-menth-1-en-8-ol, acetate, alpha-terpineol, acetate, .alpha.-terpinyl acetate, alpha-terpineol acetate, +/--alpha-terpinyl acetate, 3-cyclohexene-1-methanol, .alpha.,.alpha.,4-trimethyl-, acetate
PubChem CID 111037
ChEBI CHEBI:32320
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-yl acetate
SMILES CC(=O)OC(C)(C)C1CCC(C)=CC1

Specifications

Color Colorless
Boiling Point 94°C
Quantity 25 mL
Formula Weight 196.29
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Terpinyl Acetate
EINECSNumber (3)-2358
RTECSNumber OT0200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.