Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Terrific Broth

Catalog No. AAH2682422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2682422 100 g
AAH2682436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2682422 Supplier Thermo Scientific Chemicals Supplier No. H2682422
Add to Cart

Terrific broth is a nutritionally rich medium for the growth of bacteria and used to promote strain maintenance, cloning, and protein expression.

Terrific Broth (TB) is used with glycerol in cultivating recombinant strains of Escherichia coli. Terrific Broth II is a rich nutrient media for propagation of Escherichia coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using Terrific Broth in comparison to traditional LB Broth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Terrific broth is a nutritional medium mainly constituted by peptone, yeast extract, glycerol (as a carbon source), amino acids, and water-soluble vitamins

Application

• Terrific broth is a highly enriched medium used with glycerol to cultivate bacteria, including Escherichia coli and recombinant strains

• Terrific broth medium is commonly used for maintainance of bacterial strains, for cloning, and for protein expression

• It is a certified medium for plasmid and other bacterial cultures in a liquid medium

• A higher cell density is obtained with this media compared to traditional LB broth

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Packaging Type Glass Bottle
Quantity 100 g
Sensitivity Hygroscopic; Moisture sensitive

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.