Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Terrific Broth II (Powder)
GSA_VA

Catalog No. MP113046052 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
15 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113046052 15 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113046052 Supplier MP Biomedicals™ Supplier No. 113046052
Add to Cart
Add to Cart

Shipped as a single powder instead of separate components. MP Biomedicals Terrific Broth II (Powder) is a rich medium for the growth of E. coli. The proprietary formula contains yeast extract, tryptone, sodium chloride and vitamins. Exhibits the same grow

Terrific Broth II is a rich nutrient media for propagation of Escherichia coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using Terrific Broth in comparison to traditional LB broth.

Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in Terrific Broth II formulations.

  • Chosen reagents are combined in the accurate proportions according to the formula and thoroughly blended for a uniform distribution
  • A sample of the blended formulation is used to prepare liquid
  • The combinations are mixed to dissolve components and autoclaved at 121°C for 15 minutes
  • It is recommended to use 50g of powder to a liter of purified water

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Quantity 15 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.