Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Terrific Broth, Modified (Granulated)
GSA_VA

Catalog No. BP97282 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP9728-2 2 kg Poly Bottle
BP9728-500 500 g Poly Bottle
BP97285 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP9728-2
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 47.6g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4 mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Greater peptone and yeast concentration allow for higher plasmid yield
  • Format: Granulated powder

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
Formulation Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.