Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terrific Broth, Modified Powder, MilliporeSigma™ Supelco™

Catalog No. 111004883 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4883 250 g
11-100-4234 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4883 Supplier MilliporeSigma Supelco Supplier No. T0918250G
Add to Cart
Add to Cart

Supplement: 8 ml/l glycerol (Cat. No. 49767)

TRUSTED_SUSTAINABILITY

Specifications

Media Type Terrific Broth
Product Type Nutrient Broth
Packaging Type Poly Bottle
Quantity 250 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.