Learn More
Description
Formulation Usage: 50.8g/L
Specifications
Specifications
| Content And Storage | 4°C |
| Form | Powder |
| Grade | Molecular genetics |
| Media Type | Terrific Broth |
| Product Type | Microbiology Media |
| For Use With (Application) | Cultivation of recombinant strains of Escherichia coli |
| Packaging Type | Poly Bottle |
| Quantity | 100 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.