Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Terrific Broth (Powder/Molecular Genetics)
GSA_VA

Catalog No. BP24682 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10,000 g
100 g
2000 g
500 g
Packaging Type:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP24682 2000 g Poly Bottle
BP2468500 500 g Poly Bottle
BP2468100 100 g Poly Bottle
BP246810 10,000 g Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP24682
  • $815.50
    $562.65 / Each

    Save $252.85

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,630.00
    $1,814.65 / Case of 4 EA

    Save $815.35

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Enriched medium used with glycerol in cultivating recombinant strains of Escherichia coli.

Formulation Usage: 50.8g/L

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Description Bacterial Culture
Form Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
Formulation Water, 94.5%; Yeast extract, 2.4%; Dipotassium phosphate, 1.3%; Trptone, 1.2%; Glycerin, 0.4%; Dihydrogen potassium phosphate, 0.2%
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Poly Bottle
Quantity 2000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.