Learn More
Thermo Scientific™ Terry Tool Tubing Cutters
Catalog No. 03052228 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
1/16 in. Stainless Steel Tubing
1/8 in. Stainless Steel Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-052-228 1/8 in. Stainless Steel Tubing
03-052-227 1/16 in. Stainless Steel Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-052-228 Supplier Thermo Scientific™ Supplier No. 60182510
Add to Cart
Add to Cart
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
TRUSTED_SUSTAINABILITY
Specifications
Type Tubing Cutter
Description Cuts 0.125 stainless steel tubing
For Use With (Equipment) 1/8 in. Stainless Steel Tubing
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.