Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%, Solstice

Catalog No. 6001640 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-016-40 1 L Glass Bottle
60-016-41 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-016-40 Supplier Solstice Supplier No. 3082501L

CHROMASOLV™, for HPLC,≥99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 23°C to 26°C (literature)
Absorbance max. 0.01 at 350 nm, max. 0.01 at 300 nm, max. 0.20 at 250 nm, max. 0.50 at 230 nm, max. 1.00 at 215 nm
CAS Max % 100.0000%
Density 0.775 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 11°C
Packaging Glass Bottle
Linear Formula (CH3)3COH
UN Number UN1120
Beilstein 906698
Boiling Point 81°C to 83°C
Formula Weight 74.12
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Butanols
EINECSNumber 200-889-7
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.