Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tert-Butanol, puriss. p.a. ACS Reagent, ≥99.7% (GC), Solstice

Catalog No. 60045077 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-077 1 L Poly Bottle
60-045-079 500 mL Poly Bottle
60-045-078 2.5 L Poly Bottle
60-045-080 5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-045-077 Supplier Solstice Supplier No. 194601L
Add to Cart
Add to Cart

Puriss. p.a., ACS reagent,≥99.7% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25°C
Color Colorless
Density 0.775g/mL at 25°C
Quantity 1 L
Explosion Limit 0.08
Flash Point 11°C (51.8°F)
Ignition Point 896°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.387(lit.)
Linear Formula (CH3)3COH
Vapor Pressure 44mmHg (26°C), 31mmHg (20°C)
UN Number UN1120
Beilstein 906698
Boiling Point 83°C
Formula Weight 74.12g/mol
Percent Purity ≥99.7% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Tert-Butanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H332-H335-H336
DOTInformation Class 3 - PG 2 - Butanols
EINECSNumber 200-889-7
RTECSNumber EO1925000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.