Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (2R,3R)-3-amino-2-hydroxybutanoate 97%, Thermo Scientific™

Catalog No. p-81136
Click to view available options
Quantity:
100 mg
250 mg

Catalog No. AC338031000

Supplier: Thermo Scientific Chemicals 338031000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 204587-91-7
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
InChI Key KMUQLNSVLPPTEO-PHDIDXHHSA-N
Synonym tert-butyl 2r,3r-3-amino-2-hydroxybutanoate, butanoic acid,3-amino-2-hydroxy-, 1,1-dimethylethyl ester, 2r,3r, 2r,3r-tert-butyl 3-amino-2-hydroxybutanoate, tert-butyl 2>r,3r-3-amino-2-hydroxybutanoate, tert-butyl-2r,3r-3-amino-2-hydroxybutanoate
PubChem CID 2733830
IUPAC Name tert-butyl (2R,3R)-3-amino-2-hydroxybutanoate
SMILES CC(C(C(=O)OC(C)(C)C)O)N

Specifications

Quantity 100 mg
Formula Weight 175.23
Chemical Name or Material tert-Butyl (2R, 3R)-3-amino-2-hydroxybutanoate, 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.