Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (2R,3S)-3-amino-2-hydroxy-4-phenylbutanoate 97%, Thermo Scientific™

Catalog No. AC338001000
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338001000 100 mg
AC338002500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC338001000 Supplier Thermo Scientific Chemicals Supplier No. 338001000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119626-06-1
Molecular Formula C14H22NO3+
Molecular Weight (g/mol) 252.334
InChI Key ITPFEPDHSLWUED-NWDGAFQWSA-O
Synonym 1r,2s-1-hydroxy-3-phenyl-1-tert-butoxycarbonyl-propan-2-yl azanium, 2s,3r-4-tert-butoxy-3-hydroxy-4-oxo-1-phenylbutan-2-aminium, 2s,3r-3-hydroxy-4-2-methylpropan-2-yl oxy-4-oxo-1-phenylbutan-2-yl azanium
PubChem CID 7021088
IUPAC Name [(2S,3R)-3-hydroxy-4-[(2-methylpropan-2-yl)oxy]-4-oxo-1-phenylbutan-2-yl]azanium
SMILES CC(C)(C)OC(=O)C(C(CC1=CC=CC=C1)[NH3+])O

Specifications

Quantity 100 mg
Formula Weight 251.32
Chemical Name or Material tert-Butyl (2R, 3S)-3-amino-2-hydroxy-4-phenylbutanoate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.