Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate 98.0+%, TCI America™
SDP

Catalog No. B52481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52481G 1 g
B52485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52481G Supplier TCI America Supplier No. B52481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122684-34-8
Molecular Formula C10H18N2O3
Molecular Weight (g/mol) 214.265
MDL Number MFCD04116210
InChI Key NHDGOVOBEZPXMY-UHFFFAOYSA-N
Synonym 3-aminocarbonyl-1-boc-pyrrolidine, 1-boc-3-carbamoylpyrrolidine, 1-boc-pyrrolidine-3-carboxamide, 3-carbamoyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-pyrrolidine-3-carboxyamide, tert-butyl 3-aminocarbonyl pyrrolidine-1-carboxylate, acmc-20a2cc, ksc496a4t, 1-boc-3-pyrrolidinecarboxamide, 1-n-boc-pyrrolidine-3-carboxyamide
PubChem CID 4571096
IUPAC Name tert-butyl 3-carbamoylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)N

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 1 g
Formula Weight 214.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 3-Carbamoylpyrrolidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.