Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™

Catalog No. CC30713DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC30713DA 1 g Amber Glass Bottle
CC30713CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC30713DA Supplier Thermo Scientific Chemicals Supplier No. CC30713DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 174855-53-9
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.395
MDL Number MFCD04115066
InChI Key UINNZXQKTNAOBL-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminomethyl phenyl piperazine-1-carboxylate, 1-boc-4-2-aminomethyl phenyl piperazine, 1-boc-4-2-aminomethyl-phenyl-piperazine, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid, 4-2-aminomethyl phenyl-, 1,1-dimethylethyl ester, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic a, 2-4-t-butoxycarbonylpiperazin-1-yl phenylmethylamine, 4-2-aminomethyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminomethyl phenyl-1-piperazinecarboxylic acid tert-butyl ester
PubChem CID 4961274
IUPAC Name tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.