Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(4-formylbenzyl)piperazine-1-carboxylate, 95%, Thermo Scientific™

Catalog No. CC30304CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC30304CB 250 mg
CC30304DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC30304CB Supplier Thermo Scientific Chemicals Supplier No. CC30304CB
Add to Cart

Chemical Identifiers

CAS 844891-09-4
Molecular Formula C17H24N2O3
Molecular Weight (g/mol) 304.39
InChI Key UMXKYTSTYWRBBK-UHFFFAOYSA-N
Synonym tert-butyl 4-4-formylbenzyl piperazine-1-carboxylate, tert-butyl 4-4-formylphenyl methyl piperazine-1-carboxylate, 4-piperazin-4-yl methyl benzaldehyde, n1-boc protected, tert-butyl 4-4-formylbenzyl-1-piperazinecarboxylate, tert-butyl-4-4-formylbenzyl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-4-formylphenyl methyl-, 1,1-dimethylethyl ester
PubChem CID 2795517
IUPAC Name tert-butyl 4-[(4-formylphenyl)methyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 304.39
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(4-formylbenzyl)piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.